Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITM2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587859
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587859 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587859 Supplier Invitrogen™ Supplier No. PA587859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVKIAFNTPT AVQKEEARQD VEALLSRTVR TQILTGKELR VATQEKEGSS GRCMLTLLGL SFILAGLIVG GACIYKYFMP KSTIYRGEMC FFDSEDPANS LRGGEPNFLP VTEEADIRED DNIAIIDVPV PSFSDSDPAA IIHDFEKGMT AYLDLLLGNC YLMPLNTSIV MPPKNLVELF GKLASGRYLP QTYVVREDLV AVEEIRDVSN LGIFIYQLCN NRKSFRLRRR DLLLGFNKRA IDKCWKIRHF PNEFIVETKI CQE; Positive Samples: Mouse brain, Mouse lung, Rat brain; Cellular Location: Membrane, Single-pass type II membrane protein.

This gene encodes a type II membrane protein that belongs to the ITM2 family. Studies in mouse suggest that it may be involved in osteo- and chondrogenic differentiation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITM2A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ITM2A
Gene Accession No. Q61500
Gene Alias AW108051; BRICD2A; BRICHOS domain containing 2A; E25; E25A; Integral membrane protein 2A; Itm2; ITM2A; Protein E25; UNQ603/PRO1189
Gene Symbols ITM2A
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-263 of human ITM2A (NP_0048581).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16431, 317218
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.