Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITM2B Antibody (OTI1C11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271460F
Description
ITM2B Monoclonal specifically detects ITM2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
ITM2B | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ITM2B | |
Full length recombinant protein of human ITM2B (NP_068839) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry, Immunohistochemistry | |
OTI1C11 | |
Flow Cytometry, Immunohistochemistry | |
ABRI, ABri/ADan amyloid peptide, BRI2, BRICD2B, BRICHOS domain containing 2B, BRIFBD, E25B, E3-16, integral membrane protein 2B, Protein E25B, Transmembrane protein BRI | |
Mouse | |
Affinity Purified | |
Neuroscience, Signal Transduction | |
9445.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction