Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITM2B Monoclonal Antibody (OTI1C11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502734S


Description
ITM2B Monoclonal antibody specifically detects ITM2B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
ITM2B | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96JY6 | |
Mouse | |
Affinity chromatography | |
RUO | |
64236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C11 | |
Unconjugated | |
PDLIM2 | |
ABRI; ABri/ADan amyloid peptide; ABri23; AI256040; Bri; BRI2; BRI2 ICD; BRI2 intracellular domain; BRI2, membrane form; Bri2-23; Bri23 peptide; BRI2C, soluble form; Bricd2b; BRICHOS domain containing 2B; C-terminal peptide; D14Sel6; E25B; E25BMM; E3-16; FBD; imBRI2; Immature BRI2; integral membrane protein 2B; Integral membrane protein 2B (E25B protein); ITM2B; Mature BRI2; mBRI2; OTTHUMP00000018394; P23 peptide; Protein E25B; RDGCA; RP11-196P14.1; Transmembrane protein BRI | |
Full length human recombinant protein of human PDLIM2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction