Learn More
Invitrogen™ ITM2B Polyclonal Antibody

Description
Immunogen sequence: YFALQPDDVY YCGIKYIKDD VILNEPSADA PAALYQTIEE NIKIFEEEEV EFISVPVPEF ADSDPANIVH DFNKKLTAYL DLNLDKCYVI PLNTSIVMPP RNLLELLINI KAGTYLPQSY LIHEHMVITD RIENIDHLGF FIYRLCH Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 96%.
Specifications
Specifications
| Antigen | ITM2B |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ITM2B |
| Gene Accession No. | Q9Y287 |
| Gene Alias | ABRI; ABri/ADan amyloid peptide; ABri23; AI256040; Bri; BRI2; BRI2 ICD; BRI2 intracellular domain; BRI2, membrane form; Bri2-23; Bri23 peptide; BRI2C, soluble form; Bricd2b; BRICHOS domain containing 2B; C-terminal peptide; D14Sel6; E25B; E25BMM; E3-16; FBD; imBRI2; Immature BRI2; integral membrane protein 2B; Integral membrane protein 2B (E25B protein); ITM2B; Mature BRI2; mBRI2; OTTHUMP00000018394; P23 peptide; Protein E25B; RDGCA; RP11-196P14.1; Transmembrane protein BRI |
| Gene Symbols | ITM2B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.