Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITM2C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530838
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (95%), Rat (97%), Pig (98%), Chicken (81%), Sheep (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.
Specifications
| ITM2C | |
| Polyclonal | |
| Unconjugated | |
| ITM2C | |
| 3110038L02Rik; BRI3; Bricd2c; BRICHOS domain containing 2C; cerebral protein 14; CT-BRI3; E25; E25C; hRPC.1050_D_4; hucep-14; Integral membrane protein 2C; integral membrane protein 3; Itm2c; ITM3; NPD018; PSEC0047; Transmembrane protein BRI3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81618 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NQX7 | |
| ITM2C | |
| Recombinant protein encompassing a sequence within the center region of human ITM2C. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction