Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITM2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116254
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116254 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116254 Supplier Invitrogen™ Supplier No. PA5116254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ITM2C Polyclonal Antibody for Western Blot

Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITM2C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ITM2C
Gene Accession No. Q9NQX7
Gene Alias 3110038L02Rik; BRI3; Bricd2c; BRICHOS domain containing 2C; cerebral protein 14; CT-BRI3; E25; E25C; hRPC.1050_D_4; hucep-14; Integral membrane protein 2C; integral membrane protein 3; Itm2c; ITM3; NPD018; PSEC0047; Transmembrane protein BRI3
Gene Symbols ITM2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 27-55 amino acids from the N-terminal region of human ITM2C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.