Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITM2C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981727
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1727 Supplier Bioss Supplier No. BS11859R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Negative regulator of beta amyloid peptide production. May inhibit the processing of APP by blocking its access to alpha-and beta-secretase. Binding to the beta-secretase-cleaved APP C-terminal fragment is negligible, suggesting that ITM2C is a poor gamma-secretase cleavage inhibitor. May play a role in TNF-induced cell death and neuronal differentiation.

Specifications

Antigen ITM2C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ITM2C
Gene Accession No. Q9NQX7
Gene Alias BRI3; BRICD2C; BRICHOS domain containing 2C; Cerebral protein 14; CT-BRI3; E25; E25C; hRPC.1050_D_4; hucep-14; Integral membrane protein 2C; integral membrane protein 3; ITM2C; ITM3; NPD018; PSEC0047; Transmembrane protein BRI3
Gene Symbols ITM2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ITM2C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81618
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.