Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITM2C Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30453920UL
Description
ITM2C Polyclonal specifically detects ITM2C in Human samples. It is validated for Western Blot.Specifications
ITM2C | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
81618 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
BRI3E25C, BRICD2C, BRICHOS domain containing 2C, Cerebral protein 14, E25, hRPC.1050_D_4, integral membrane protein 2C, integral membrane protein 3, ITM3, Transmembrane protein BRI3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ITM2C (NP_001012532.1). | |
20 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction