Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528483
Description
Recommended positive controls: 293T, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (91%), Rat (92%), Xenopus laevis (80%), Chicken (81%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pyrophosphatase that hydrolyzes the non-canonical purine nucleotides inosine triphosphate (ITP), deoxyinosine triphosphate (dITP) as well as 2'-deoxy-N-6-hydroxylaminopurine triposphate (dHAPTP) and xanthosine 5'-triphosphate (XTP) to their respective monophosphate derivatives. The enzyme does not distinguish between the deoxy- and ribose forms. Probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions.
Specifications
| ITPA | |
| Polyclonal | |
| Unconjugated | |
| ITPA | |
| 2010016I08Rik; AU020102; C20orf37; dJ794I6.3; HAMAP-Rule:MF_03148}; HLC14-06-P; inosine triphosphatase; inosine triphosphatase (nucleoside triphosphate pyrophosphatase); inosine triphosphatase {ECO:0000255; inosine triphosphate pyrophosphatase; inosine triphosphate pyrophosphatase {ECO:0000255; inosine triphosphate pyrophosphohydrolase; Itp; ITPA; ITPase; ITPase {ECO:0000255; My049; My049 protein; Non-canonical purine NTP pyrophosphatase; non-canonical purine NTP pyrophosphatase {ECO:0000255; non-standard purine NTP pyrophosphatase; non-standard purine NTP pyrophosphatase {ECO:0000255; NTPase; NTPase {ECO:0000255; nucleoside triphosphate pyrophosphatase; nucleoside-triphosphate diphosphatase; nucleoside-triphosphate diphosphatase {ECO:0000255; Nucleoside-triphosphate pyrophosphatase; nucleoside-triphosphate pyrophosphatase {ECO:0000255; OK/SW-cl.9; Putative oncogene protein hlc14-06-p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3704 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BY32 | |
| ITPA | |
| Recombinant protein encompassing a sequence within the center region of human ITPase. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction