Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575999 Supplier Invitrogen™ Supplier No. PA575999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Pyrophosphatase that hydrolyzes the non-canonical purine nucleotides inosine triphosphate (ITP), deoxyinosine triphosphate (dITP) as well as 2'-deoxy-N-6-hydroxylaminopurine triposphate (dHAPTP) and xanthosine 5'-triphosphate (XTP) to their respective monophosphate derivatives. The enzyme does not distinguish between the deoxy- and ribose forms. Probably excludes non-canonical purines from RNA and DNA precursor pools, thus preventing their incorporation into RNA and DNA and avoiding chromosomal lesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ITPA
Gene Accession No. D3ZW55, Q9BY32, Q9D892
Gene Alias 2010016I08Rik; AU020102; C20orf37; dJ794I6.3; HAMAP-Rule:MF_03148}; HLC14-06-P; inosine triphosphatase; inosine triphosphatase (nucleoside triphosphate pyrophosphatase); inosine triphosphatase {ECO:0000255; inosine triphosphate pyrophosphatase; inosine triphosphate pyrophosphatase {ECO:0000255; inosine triphosphate pyrophosphohydrolase; Itp; ITPA; ITPase; ITPase {ECO:0000255; My049; My049 protein; Non-canonical purine NTP pyrophosphatase; non-canonical purine NTP pyrophosphatase {ECO:0000255; non-standard purine NTP pyrophosphatase; non-standard purine NTP pyrophosphatase {ECO:0000255; NTPase; NTPase {ECO:0000255; nucleoside triphosphate pyrophosphatase; nucleoside-triphosphate diphosphatase; nucleoside-triphosphate diphosphatase {ECO:0000255; Nucleoside-triphosphate pyrophosphatase; nucleoside-triphosphate pyrophosphatase {ECO:0000255; OK/SW-cl.9; Putative oncogene protein hlc14-06-p
Gene Symbols ITPA
Host Species Rabbit
Immunogen Recombinant full length Human ITPA.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16434, 311422, 3704
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.