Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITPK1 Antibody (OTI1F5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271052F
Description
ITPK1 Monoclonal specifically detects ITPK1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
ITPK1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ITPK1 | |
Recombinant protein fragment corresponding to amino acids 309-391 of human ITPK1 (NP_001136065) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI1F5 | |
Immunohistochemistry | |
EC 2.7.1.134, EC 2.7.1.159, inositol 13,4-triphosphate 5/6 kinase, inositol 13,4-trisphosphate 5/6 kinase, Inositol 13,4-trisphosphate 5/6-kinase, inositol-tetrakisphosphate 1-kinase, Inositol-triphosphate 5/6-kinase, Ins(13,4)P(3) 5/6-kinase, ITRPK1 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
3705.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction