Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITPK1 Antibody (SR1711), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322570100UL
Description
ITPK1 Monoclonal antibody specifically detects ITPK1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ITPK1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1711 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
EC 2.7.1.134, EC 2.7.1.159, inositol 13,4-triphosphate 5/6 kinase, inositol 13,4-trisphosphate 5/6 kinase, Inositol 13,4-trisphosphate 5/6-kinase, inositol-tetrakisphosphate 1-kinase, Inositol-triphosphate 5/6-kinase, Ins(13,4)P(3) 5/6-kinase, ITRPK1 | |
A synthesized peptide derived from human ITPK1 (Uniprot #: Q13572) | |
100 μg | |
Signal Transduction | |
3705 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction