Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597001 Supplier Invitrogen™ Supplier No. PA597001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CVVQNFINHN AVLYKVFVVG ESYTVVQRPS LKNFSAGTSD RESIFFNSHN VSKPESSSVL TELDKIEGVF ERPSDEVIRE LSRALRQALG VSLFGIDIII NNQTGQHAVI DINAFPGYEG VSEFFTDLLN HIATVLQGQS TAMAATGDVA LLRHSKLLAE PAGGLVGERT CSASPGCCGS MMGQDAPWKA EADAGGTAKL PHQRLGCNAG VSPSFQQHCV ASLATKASSQ; Positive Samples: HeLa, HepG2, Mouse testis, Mouse brain, Mouse intestine, Rat brain.

ITPK1 encodes an enzyme that belongs to the inositol 1,3,4-trisphosphate 5/6-kinase family. This enzyme regulates the synthesis of inositol tetraphosphate, and downstream products, inositol pentakisphosphate and inositol hexakisphosphate. Inositol metabolism plays a role in the development of the neural tube. Disruptions in this gene are thought to be associated with neural tube defects. A pseudogene of this gene has been identified on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPK1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ITPK1
Gene Accession No. Q13572, Q8BYN3
Gene Alias BC031182; inositol 1,3,4-triphosphate 5/6 kinase; inositol 1,3,4-trisphosphate 5/6-kinase; Inositol-tetrakisphosphate 1-kinase; inositol-triphosphate 5/6-kinase; ins(1,3,4)P(3) 5/6-kinase; ITPK1; ITRPK1
Gene Symbols ITPK1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 185-414 of human ITPK1 (NP_0550312).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217837, 3705, 500709
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.