Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITPKA Polyclonal Antibody
ITPKA Polyclonal Antibody
Supplier: Thermo Scientific PA5101093
Description
ITPKA Polyclonal Antibody for Western Blot, ICC/IF

Specifications
ITPKA | |
Polyclonal | |
Unconjugated | |
ITPKA | |
EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-3KA; IP3K A; IP3KA; IP3-kinase A; ITPKA | |
Rabbit | |
Affinity Chromatography | |
RUO | |
228550, 3706, 81677 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P17105, P23677, Q8R071 | |
ITPKA | |
A synthesized peptide derived from human ITPKA(Accession P23677), corresponding to amino acid residues L37-E87. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction