Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPKA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578659 Supplier Invitrogen™ Supplier No. PA578659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse brain, rat brain Predicted Reactivity: Human (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPKA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ITPKA
Gene Accession No. P17105, P23677, Q8R071
Gene Alias EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-3KA; IP3K A; IP3KA; IP3-kinase A; ITPKA
Gene Symbols ITPKA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IP3KA. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228550, 3706, 81677
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.