Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITPKA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585786
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%). Positive Control: mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Regulates inositol phosphate metabolism by phosphorylation of second messenger inositol 1,4,5-trisphosphate to Ins(1,3,4,5)P4. The activity of the inositol 1,4,5-trisphosphate 3-kinase is responsible for regulating the levels of a large number of inositol polyphosphates that are important in cellular signaling. Both calcium/calmodulin and protein phosphorylation mechanisms control its activity. It is also a substrate for the cyclic AMP-dependent protein kinase, calcium/calmodulin- dependent protein kinase II, and protein kinase C in vitro.
Specifications
ITPKA | |
Polyclonal | |
Unconjugated | |
ITPKA | |
EC 2.7.1.127; inositol 1,4,5-triphosphate 3-kinase; inositol 1,4,5-trisphosphate 3-kinase A; inositol 145-triphosphate 3-kinase; inositol-trisphosphate 3-kinase A; insP 3-kinase A; IP3 3 kinase A; IP3 3-kinase A; IP3-3KA; IP3K A; IP3KA; IP3-kinase A; ITPKA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228550, 81677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
1.78 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P17105, Q8R071 | |
ITPKA | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse IP3KA. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction