Learn More
Invitrogen™ ITPKB Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120876
Description
Positive test controls include: HeLa, HepG2, BT-474, Mouse lung. Immunogen sequence: MAVYCYALNS LVIMNSANEM KSGGGPGPSG SETPPPPRRA VLSPGSVFSP GRGASFLFPP AESLSPEEPR SPGGWRSGRR RLNSSSGSGS GSSGSSVSSP SWAGRLRGDR QQVVAAGTLS PPGPEEAKRK LRILQRELQN VQVNQKVGMF EAHIQAQSSA IQAPRSPRLG RARSPSPCPF RSSSQPPGRV LVQGARSEER RTKSWGEQCP ETSGTDSGRK GGPSLCSSQV KKGMPPLPGR AAPTGSEAQG PSAFVRMEKG.
Inositol 1,4,5-trisphosphate (Ins(1,4,5)P3) regulates the level of calcium within the cell by releasing calcium from intracellular stores. Ins(1,4,5)P3 is phosphorylated by inositol 1,4,5-trisphosphate 3-kinase (IP3K) to form inositol 1,3,4,5-tetrakisphosphate (Ins(1,4,5)P4), which is is thought to regulate the influx of calcium across the plasma membrane. IP3K exists as three isoforms, IP3KA, B, and C. IP3KA, the most highly characterized isoform, is expressed in rat brain and testis. IP3KB is expressed in various rat tissues such as lung, thymus, testis, brain, and heart. IP3K activity is stimulated in the presence of calmodulin via phosphorylation by cAMP-dependent protein kinase, protein kinase C, or calcium/calmodulin dependent protein kinase II and, subsequently, mediates the inositol phosphate signaling pathways.
Specifications
| ITPKB | |
| Polyclonal | |
| Unconjugated | |
| ITPKB | |
| 1110033J02Rik; E130307H12Rik; Inositol 1,4,5-trisphosphate 3-kinase B; inositol-trisphosphate 3-kinase B; insP 3-kinase B; IP3 3-kinase B; IP3-3KB; IP3K; IP3K B; IP3KB; IP3K-B; Itpkb; PIG37; proliferation-inducing protein 37 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 320404, 3707, 54260 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.11 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27987, P42335 | |
| ITPKB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ITPKB (NP_002212.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.