Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPKC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111321 Supplier Invitrogen™ Supplier No. PA5111321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSWADNLWTH QNSSSLQTHP EGACPSKEPS ADGSWKELYT DGSRTQQDIE GPWTEPYTDG SQKKQDTEAA RKQ Highest antigen sequence indentity to the following orthologs: Mouse - 51%, Rat - 55%.

Inositol 1, 4, 5-trisphosphate 3-kinase (ITPK) catalyzes the phosphorylation of Ins(1, 4, 5)P3 to Ins(1, 3, 4, 5)P4, both of which are modulators of calcium homeostasis. ITPK isoforms include ITPKA, ITPKB, and ITPKC, all of which contain a conserved catalytic unit in their C termini, but have unique N-terminal sequences and tissue distributions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPKC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITPKC
Gene Accession No. Q96DU7
Gene Alias inositol 1,4,5-trisphosphate 3-kinase C; Inositol-trisphosphate 3-kinase C; InsP 3 kinase C; insP 3-kinase C; IP3 3-kinase C; IP3-3KC; IP3K C; IP3KC; ITPKC
Gene Symbols ITPKC
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ITPKC. Recombinant protein control fragment (Product #RP-91720).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.