Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITPR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ITPR2 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ITPR2 Polyclonal specifically detects ITPR2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ITPR2 | |
Polyclonal | |
Rabbit | |
Neuroscience | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
3709 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LICKFMLSPGNADILIQTKVVSMQADNPMESSILSDDIDDEEVWLYWIDSNKEPHGKAIRHLAQEAKEGTKADLEVLT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
inositol 14,5-triphosphate receptor, type 2, inositol 14,5-trisphosphate receptor type 2, insP3R2, IP3 receptor, IP3 receptor isoform 2, IP3R 2, IP3R2, Type 2 inositol 14,5-trisphosphate receptor, Type 2 InsP3 receptor | |
ITPR2 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title