Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550688 Supplier Invitrogen™ Supplier No. PA550688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total ITPR3 protein.

This gene encodes a receptor for inositol 1,4,5-trisphosphate, a second messenger that mediates the release of intracellular calcium. The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus. Knockout studies in mice suggest that type 2 and type 3 inositol 1,4,5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ITPR3
Gene Accession No. Q14573
Gene Alias inositol 1,4,5-triphosphate receptor 3; inositol 1,4,5-triphosphate receptor, type 3; inositol 1,4,5-trisphosphate receptor type 3; inositol 1,4,5-trisphosphate receptor, type 3; insP3R3; IP3 receptor; IP3 receptor isoform 3; IP3R; IP3R 3; Ip3r3; IP3R-3; IP3R3X; ITPR3; Itpr-3; Type 3 inositol 1,4,5-trisphosphate receptor; type 3 InsP3 receptor
Gene Symbols ITPR3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human inositol 1,4,5-trisphosphate receptor, type 3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3710
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.