Learn More
Invitrogen™ ITPR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588758
Description
Immunogen sequence: EGSEAYQRCE SGGFLSKLIQ HTKDLMESEE KLCIKVLRTL QQMLLKKTKY GDRGNQLRKM LLQNYLQNRK STSRGDLPDP IGTGLDPDWS AIAATQCRLD KEGATKLVCD LITSTKNEKI FQESIGLAIH LLDGGNTEIQ KSFHNLMMSD KKSERFFKVL HDRMKRAQQE TKSTVAVNMN DLGSQPHEDR EPVDPTTKGR VASFSIPGSS SRYSLGPSLR RGHEVSERVQ SSEMGTSVLI M; Positive Samples: HeLa, LO2, HepG2; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein.
This gene encodes a receptor for inositol 1,4,5-trisphosphate, a second messenger that mediates the release of intracellular calcium. The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus. Knockout studies in mice suggest that type 2 and type 3 inositol 1,4,5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth.
Specifications
ITPR3 | |
Polyclonal | |
Unconjugated | |
ITPR3 | |
inositol 1,4,5-triphosphate receptor 3; inositol 1,4,5-triphosphate receptor, type 3; inositol 1,4,5-trisphosphate receptor type 3; inositol 1,4,5-trisphosphate receptor, type 3; insP3R3; IP3 receptor; IP3 receptor isoform 3; IP3R; IP3R 3; Ip3r3; IP3R-3; IP3R3X; ITPR3; Itpr-3; Type 3 inositol 1,4,5-trisphosphate receptor; type 3 InsP3 receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16440, 3710 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.69 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P70227, Q14573 | |
ITPR3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1630-1870 of human ITPR3 (NP_0022152). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.