Learn More
ITPRIP Polyclonal Antibody

Description
ITPRIP Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | ITPRIP |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | ITPRIP |
| Gene Accession No. | Q8IWB1 |
| Gene Alias | 4833424O12Rik; bA127L20; bA127L20.2; DANGER; inositol 1,4,5-triphosphate receptor interacting protein; inositol 1,4,5-triphosphate receptor-interacting protein; inositol 1,4,5-trisphosphate receptor interacting protein; inositol 1,4,5-trisphosphate receptor-interacting protein; ipri; Itprip; KIAA1754; mKIAA1754; Protein DANGER |
| Gene Symbols | ITPRIP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.