Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPRIPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531770 Supplier Invitrogen™ Supplier No. PA531770
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (90%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The ITPRIPL1 (Inositol 1,4,5-Trisphosphate Receptor Interacting Protein-Like 1) gene encodes a protein implicated in T cell regulation by interacting with the CD3ε molecule. It is located on chromosome 6. The ITPRIPL1 protein acts as an inhibitory ligand for CD3ε, thereby modulating T cell activation in the tumor microenvironment. This interaction significantly reduces calcium influx and phosphorylation of ZAP70, which are critical steps in the T cell activation pathway. The downregulation of these processes by ITPRIPL1 suggests its role in immune response modulation and tumor immune evasion. Given its regulatory functions, ITPRIPL1 has become a subject of research for its potential implications in cancer immunotherapy and as a biomarker for diagnosing and prognosticating various cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPRIPL1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ITPRIPL1
Gene Accession No. Q6GPH6
Gene Alias inositol 1,4,5-triphosphate receptor-interacting protein-like 1; inositol 1,4,5-trisphosphate receptor interacting protein-like 1; inositol 1,4,5-trisphosphate receptor-interacting protein-like 1; ITPRIPL1; KIAA1754L
Gene Symbols ITPRIPL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human ITPRIPL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150771
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.