Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ITPRIPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565323 Supplier Invitrogen™ Supplier No. PA565323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WLRDCKPFADA FCVDVRGRRH LSATLVLRWF QSHLQRSLAT VRYSLEGRCR VTLTPGGLEQ PPTLHILPCR TDYGCCRLSM AVRLIPAVH Highest antigen sequence identity to the following orthologs - mouse 92%, rat 24%.

ITPRIPL2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ITPRIPL2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ITPRIPL2
Gene Accession No. Q3MIP1
Gene Alias inositol 1,4,5-triphosphate receptor interacting protein-like 2; inositol 1,4,5-triphosphate receptor-interacting protein-like 2; inositol 1,4,5-trisphosphate receptor interacting protein-like 2; inositol 1,4,5-trisphosphate receptor-interacting protein-like 2; ITPRIPL2
Gene Symbols ITPRIPL2
Host Species Rabbit
Immunogen Recombinant Human ITPRIPL2. Recombinant protein control fragment (Product #RP-98807).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.