Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITSN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598440
Description
ITSN1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a cytoplasmic membrane-associated protein that indirectly coordinates endocytic membrane traffic with the actin assembly machinery. In addition, the encoded protein may regulate the formation of clathrin-coated vesicles and could be involved in synaptic vesicle recycling. This protein has been shown to interact with dynamin, CDC42, SNAP23, SNAP25, SPIN90, EPS15, EPN1, EPN2, and STN2. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been characterized so far.
Specifications
ITSN1 | |
Polyclonal | |
Unconjugated | |
ITSN1 | |
AA517634; AA545208; AI316805; AI839402; AI848451; EH and SH3 domains protein 1; EH domain and SH3 domain regulator of endocytosis 1; Eh domain, SH3 domain regulator of endocytosis 1; Ehsh1; Ese1; human intersectin-SH3 domain-containing protein SH3P17; intersectin (SH3 domain protein 1A); intersectin 1; intersectin 1 (SH3 domain protein 1A); intersectin 1 (SH3 domain protein); intersectin 1 short form variant 3; intersectin 1 short form variant, 11; intersectin 1; intersectin-1; intersectin short form 2 variant 2; intersectin short variant 12; intersectin-1; intersectin-L; Itsn; ITSN1; MGC134948; MGC134949; SH3 domain-containing protein 1A; SH3D1A; SH3P17; Src homology 3 domain-containing protein | |
Rabbit | |
Protein G | |
RUO | |
6453 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.775 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q15811 | |
ITSN1 | |
Recombinant Human Intersectin-1 protein (773-916AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction