Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ITSN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115432
Description
Antibody detects endogenous levels of total Intersectin 1.
The protein encoded by this gene is a cytoplasmic membrane-associated protein that indirectly coordinates endocytic membrane traffic with the actin assembly machinery. In addition, the encoded protein may regulate the formation of clathrin-coated vesicles and could be involved in synaptic vesicle recycling. This protein has been shown to interact with dynamin, CDC42, SNAP23, SNAP25, SPIN90, EPS15, EPN1, EPN2, and STN2. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been characterized so far.
Specifications
| ITSN1 | |
| Polyclonal | |
| Unconjugated | |
| ITSN1 | |
| AA517634; AA545208; AI316805; AI839402; AI848451; EH and SH3 domains protein 1; EH domain and SH3 domain regulator of endocytosis 1; Eh domain, SH3 domain regulator of endocytosis 1; Ehsh1; Ese1; human intersectin-SH3 domain-containing protein SH3P17; intersectin (SH3 domain protein 1A); intersectin 1; intersectin 1 (SH3 domain protein 1A); intersectin 1 (SH3 domain protein); intersectin 1 short form variant 3; intersectin 1 short form variant, 11; intersectin 1; intersectin-1; intersectin short form 2 variant 2; intersectin short variant 12; intersectin-1; intersectin-L; Itsn; ITSN1; MGC134948; MGC134949; SH3 domain-containing protein 1A; SH3D1A; SH3P17; Src homology 3 domain-containing protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16443, 29491, 6453 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q15811, Q9WVE9, Q9Z0R4 | |
| ITSN1 | |
| A synthesized peptide derived from human ITSN1(Accession Q15811), corresponding to amino acid residues L274-S324. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction