Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ivacaftor
GREENER_CHOICESDP

Catalog No. 788750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-875-0 50 mg
78-871-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 78-875-0 Supplier Tocris Bioscience™ Supplier No. 7887/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CFTR potentiator

Ivacaftor is a potent and selective potentiator of Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) that targets F508del-CFTR and G551D-CFTR (EC50 values are 25 nM and 100 nM, respectively); its potentiation activity depends on the level of CFTR phosphorylation. Ivacaftor reversibly binds to wild type (WT-CFTR) and G551D-CFTR mutants, increasing the open probability via an ATP-independent mechanism, and enhances cAMP/PKA-signaling mediated gating activity for both WT-CFTR and G551D-CFTR mutants. In cultured human CF bronchial epithelia with G551D/F508del mutations, Ivacaftor increases Forskolin (Cat. No. 1099)-induced transepithelial current (EC50 = 236 nM) and increases chloride secretion by 10-fold. Also rescues endothelial CFTR expression and function and prevents endothelial barrier failure and protein leak in human pulmonary microvascular endothelial cells. Ivacaftor is orally bioavailable.

Specifications

Chemical Name or Material N-[2,4-Bis(1,1-dimethylethyl)-5-hydroxyphenyl]-1,4-dihydro-4-oxo-3-quinolinecarboxamide
CAS 873054-44-5
Quantity 50 mg
Target CFTR
Molecular Formula C24H28N2O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.