Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IVD Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318798JF549
Description
IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for ELISASpecifications
IVD | |
Polyclonal | |
ELISA | |
ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial | |
Produced in rabbits immunized with purified recombinant Human IVD (His30-His423, Accession: AAH17202.1) | |
0.1 mL | |
Stem Cell Markers | |
3712 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction