Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IVD Antibody (OTI1B10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271055H
Description
IVD Monoclonal specifically detects IVD in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
IVD | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
IVD | |
Full length recombinant protein of human IVD (NP_002612) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1B10 | |
Western Blot, Immunohistochemistry | |
ACAD2, EC 1.3.99, EC 1.3.99.10, FLJ12715, FLJ34849, isovaleryl-CoA dehydrogenase, mitochondrial | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
3712.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction