Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IVD Monoclonal Antibody (OTI1B10)
IVD Monoclonal Antibody (OTI1B10)
Supplier: Thermo Scientific MA525246
Description
IVD Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
IVD | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12007, P26440 | |
IVD | |
Full length human recombinant protein of IVD produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Rat, Canine, Monkey | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI1B10 | |
Unconjugated | |
IVD | |
1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; butyryl-CoA dehydrogenase; EGK_17362; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; isovaleryl-CoA dehydrogenase, mitochondrial; IVD | |
Mouse | |
Affinity Chromatography | |
RUO | |
100856316, 24513, 3712 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction