Learn More
IVD Mouse anti-Canine, Human, Rat, Clone: OTI1B10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501742

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Isovaleryl-CoA dehydrogenase (IVD) is a mitochondrial matrix enzyme that catalyzes the third step in leucine catabolism. The genetic deficiency of IVD results in an accumulation of isovaleric acid, which is toxic to the central nervous system and leads to isovaleric acidemia.Specifications
IVD | |
Monoclonal | |
Unconjugated | |
IVD | |
ACAD2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
100856316, 24513, 3712 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P12007, P26440 | |
IVD | |
Full length human recombit protein of human IVD produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.