Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IVD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522342 Supplier Invitrogen™ Supplier No. PA522342
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, Jurkat, mouse brain, PC-12. Predicted reactivity: Mouse (90%), Rat (90%), Pig (91%), Chicken (85%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Isovaleryl-CoA dehydrogenase (IVD) is a mitochondrial matrix enzyme that catalyzes the third step in leucine catabolism. The genetic deficiency of IVD results in an accumulation of isovaleric acid, which is toxic to the central nervous system and leads to isovaleric acidemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IVD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene IVD
Gene Accession No. P12007, P26440, Q9JHI5
Gene Alias 1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; butyryl-CoA dehydrogenase; EGK_17362; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; isovaleryl-CoA dehydrogenase, mitochondrial; IVD
Gene Symbols IVD
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IVD. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24513, 3712, 56357
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.