Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IVD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559547 Supplier Invitrogen™ Supplier No. PA559547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLPVDDAING LSEEQRQLRQ TMAKFLQEHL APKAQEIDRS NEFKNLREFW KQLGNLGVLG ITAPVQYGGS GLGYLEHVLV MEEISRASGA VGLSYGAHSN LCINQLVRNG NE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Isovaleryl-CoA dehydrogenase (IVD) is a mitochondrial matrix enzyme that catalyzes the third step in leucine catabolism. The genetic deficiency of IVD results in an accumulation of isovaleric acid, which is toxic to the central nervous system and leads to isovaleric acidemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IVD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IVD
Gene Accession No. P26440
Gene Alias 1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; butyryl-CoA dehydrogenase; EGK_17362; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; isovaleryl-CoA dehydrogenase, mitochondrial; IVD
Gene Symbols IVD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IVD. Recombinant protein control fragment (Product #RP-98064).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.