Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IVD Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5112148
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112148 Supplier Thermo Scientific Supplier No. PA5112148

Rabbit Polyclonal Antibody

This product can be used in direct ELISA.

IVD Polyclonal antibody specifically detects IVD in Human samples. It is validated for Immunoprecipitation, Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IVD
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IVD
Gene Accession No. P26440
Gene Alias 1300016K07Rik; 6720455E18Rik; ACAD2; AI463340; BOS_10725; isovaleryl CoA dehydrogenase; isovaleryl coenzyme A dehydrogenase; isovaleryl dehydrogenase; isovaleryl-CoA dehydrogenase; Isovaleryl-CoA dehydrogenase, mitochondrial; Ivd
Host Species Rabbit
Immunogen Recombinant Human Isovaleryl-CoA dehydrogenase/IVD Protein
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3712
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.