Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IVL Rabbit anti-Dog, Gorilla, Human, Owl Monkey, Pig, Unconjugated, Clone: IVRN/2113R, Abnova™

Catalog No. 89337888 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-888 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-888 Supplier Abnova Corporation Supplier No. RAB00463
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

IVL Monoclonal antibody specifically detects IVL in Dog, Gorilla, Human, Owl Monkey, Pig samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen IVL
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone IVRN/2113R
Conjugate Unconjugated
Description involucrin
Gene IVL
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human IVL
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3713
Target Species Dog, Gorilla, Human, Owl Monkey, Pig
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.