Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IVL Rabbit anti-Dog, Gorilla, Human, Owl Monkey, Pig, Unconjugated, Clone: IVRN/2113R, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation RAB00464
Description
IVL Monoclonal antibody specifically detects IVL in Dog, Gorilla, Human, Owl Monkey, Pig samples. It is validated for IHC-P, IF, Flow Cyt.Specifications
IVL | |
Monoclonal | |
Unconjugated | |
IVL | |
Recombinant protein corresponding to full-length human IVL | |
Primary | |
Dog, Gorilla, Human, Owl Monkey, Pig | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry | |
IVRN/2113R | |
involucrin | |
Rabbit | |
100 μg | |
3713 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction