Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IVNS1ABP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100582 Supplier Invitrogen™ Supplier No. PA5100582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IVNS1ABP.

IVNS1ABP is a protein coding gene involved in many cell functions, including pre-mRNA splicing, the aryl hydrocarbon receptor (AHR) pathway, F-actin organization and protein ubiquitination. Plays a role in the dynamic organization of the actin skeleton as a stabilizer of actin filaments by association with F-actin through Kelch repeats. Protects cells from cell death induced by actin destabilization. Functions as modifier of the AHR/Aryl hydrocarbon receptor pathway increasing the concentration of AHR available to activate transcription. In addition, functions as a negative regulator of BCR(KLHL20) E3 ubiquitin ligase complex to prevent ubiquitin-mediated proteolysis of PML and DAPK1, two tumor suppressors. Inhibits pre-mRNA splicing (in vitro).; (Microbial infection) involved in the alternative splicing of influenza A virus M1 mRNA through interaction with HNRNPK, thereby facilitating the generation of viral M2 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IVNS1ABP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IVNS1ABP
Gene Accession No. Q920Q8, Q9Y6Y0
Gene Alias 1190004M08Rik; 1700126I16Rik; AA960440; ARA3; aryl hydrocarbon receptor-associated 3; aryl hydrocarbon receptor-associated protein 3; FLARA3; HSPC068; influenza virus NS1A binding protein; influenza virus NS1A-binding protein; Influenza virus NS1A-binding protein homolog; Ivns1abp; kelch family protein Nd1; kelch family protein Nd1-L; kelch-like family member 39; Kelch-like protein 39; KIAA0850; KLHL39; mKIAA0850; NCX downstream gene 1; ND1; Nd1L; Nd1-L; ND1-L2; Nd1S; Nd1-S; NS1; NS-1; NS1-binding protein; NS1-binding protein homolog; Ns1bp; NS1-BP
Gene Symbols IVNS1ABP
Host Species Rabbit
Immunogen A synthesized peptide derived from human IVNS1ABP(Accession Q9Y6Y0), corresponding to amino acid residues T403-R453.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10625, 117198, 289089
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.