Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ IWP 2
GREENER_CHOICESDP

Catalog No. 353310 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-331-0 10 mg
35-335-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-331-0 Supplier Tocris Bioscience™ Supplier No. 3533/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PORCN inhibitor; inhibits Wnt processing and secretion

IWP 2 is a potent inhibitor of Wnt processing and secretion (IC50 = 27nM). IWP 2 inactivates PORCN, a membrane-bound O-acyltransferase (MBOAT), and selectively inhibits palmitoylation of Wnt. Blocks Wnt-dependent phosphorylation of Lrp6 receptor and Dvl2, and β-catenin accumulation. IWP 2 suppresses self-renewal in R1 embryonic stem cells and promotes cardiomyocyte differentiation from hPSCs.

Chemical Identifiers

CAS 686770-61-6
Molecular Formula C22H18N4O2S3

Specifications

Quantity 10 mg
Formula Weight 466.6
Purity >98%
Molecular Weight (g/mol) 466.59
Chemical Name or Material IWP 2
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.