Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IWS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557374
Description
Immunogen sequence: IFGESGDEEE EEFTGFNQED LEEEKGETQV KEAEDSDSDD NIKRGKHMDF LSDFEMMLQR KKSMSGKRRR NRDGGTFISD ADDVVSAMIV KMNEAA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
Specifications
IWS1 | |
Polyclonal | |
Unconjugated | |
IWS1 | |
1700069O15Rik; 3000008H23; AL117927; AW214353; interacts with Spt6; IWS1; IWS1 homolog; IWS1 homolog (S. cerevisiae); IWS1, SUPT6H interacting protein; Iws1l; IWS1-like protein; Protein IWS1 homolog; RGD1304762 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96ST2 | |
IWS1 | |
Recombinant protein corresponding to Human IWS1. Recombinant protein control fragment (Product #RP-96178). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction