Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IWS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567630 Supplier Invitrogen™ Supplier No. PA567630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Rat samples.

Transcription factor which plays a key role in defining the composition of the RNA polymerase II (RNAPII) elongation complex and in modulating the production of mature mRNA transcripts. Acts as an assembly factor to recruit various factors to the RNAPII elongation complex and is recruited to the complex via binding to the transcription elongation factor SUPT6H bound to the C-terminal domain (CTD) of the RNAPII subunit RPB1 (POLR2A). The SUPT6H:IWS1:CTD complex recruits mRNA export factors (ALYREF/THOC4, EXOSC10) as well as histone modifying enzymes (such as SETD2) to ensure proper mRNA splicing, efficient mRNA export and elongation-coupled H3K36 methylation, a signature chromatin mark of active transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IWS1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene IWS1
Gene Accession No. Q3SWT4, Q96ST2
Gene Alias 1700069O15Rik; 3000008H23; AL117927; AW214353; interacts with Spt6; IWS1; IWS1 homolog; IWS1 homolog (S. cerevisiae); IWS1, SUPT6H interacting protein; Iws1l; IWS1-like protein; Protein IWS1 homolog; RGD1304762
Gene Symbols IWS1
Host Species Rabbit
Immunogen Synthesized peptide derived from human IWS1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291705, 55677
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.