Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IYD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563757 Supplier Invitrogen™ Supplier No. PA563757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADRSMEKKKG EPRTRAEARP WVDEDLKDSS DLHQAEEDAD EWQESEENVE HIPFSHNHYP EK Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.

This gene encodes an enzyme that catalyzes the oxidative NADPH-dependent deiodination of mono- and diiodotyrosine, which are the halogenated byproducts of thyroid hormone production. The N-terminus of the protein functions as a membrane anchor. Mutations in this gene cause congenital hypothyroidism due to dyshormonogenesis type 4, which is also referred to as deiodinase deficiency, or iodotyrosine dehalogenase deficiency, or thyroid hormonogenesis type 4. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IYD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IYD
Gene Accession No. Q6PHW0
Gene Alias C6orf71; DEHAL1; dJ422F24.1; iodotyrosine dehalogenase 1; iodotyrosine dehalogenase 1; iodotyrosine deiodinase 1; iodotyrosine deiodinase; iodotyrosine deiodinase 1; IYD; IYD-1; RGD1309288; TDH4
Gene Symbols IYD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IYD. Recombinant protein control fragment (Product #RP-101082).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.