Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IYD Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501731099 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1099 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1099 Supplier Proteintech Group Inc Supplier No. 166671AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an enzyme that catalyzes the oxidative NADPH-dependent deiodination of mono- and diiodotyrosine, which are the halogenated byproducts of thyroid hormone production. The N-terminus of the protein functions as a membrane anchor. Mutations in this gene cause congenital hypothyroidism due to dyshormonogenesis type 4, which is also referred to as deiodinase deficiency, or iodotyrosine dehalogenase deficiency, or thyroid hormonogenesis type 4. Alternative splicing results in multiple transcript variants.

Specifications

Antigen IYD
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IYD
Gene Accession No. Q6PHW0
Gene Alias C6orf71, DEHAL1, Iodotyrosine dehalogenase 1, iodotyrosine deiodinase, IYD, IYD 1
Gene Symbols IYD
Host Species Rabbit
Immunogen IYD Fusion Protein Ag10146
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389434
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.