Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IZUMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543687
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543687 Supplier Invitrogen™ Supplier No. PA543687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SLALITGLTF AIFRRRKVID FIKSSLFGLG SGAAEQTQVP KEKATDSRQQ Sequence homology: Human: 100%.

The sperm-specific protein Izumo, named for a Japanese shrine dedicated to marriage, is essential for sperm-egg plasma membrane binding and fusion.The sperm-specific protein Izumo, named for a Japanese shrine dedicated to marriage, is essential for sperm-egg plasma membrane binding and fusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IZUMO1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Izumo1
Gene Accession No. Q8IYV9
Gene Alias 1700058F15Rik; Izumo; izumo sperm-egg fusion 1; izumo sperm-egg fusion protein 1; IZUMO1; MGC34799; OBF; oocyte binding/fusion factor; Sperm-specific protein izumo
Gene Symbols Izumo1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human IZUMO1 (aa 301-350).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284359
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.