Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IZUMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558248 Supplier Invitrogen™ Supplier No. PA558248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVLALKSLEK DYLPGHLDAK HHKAMMERVE NAVKDFQELS LNEDAYMGVV DEATLQKGSW SLLKDLKRIT DSDVKGDLFV KELFWMLHLQ KETFATYVAR FQKEAYCPNK CGVMLQTLIW CKNCKKEVHA CRKSYDCGER NVEVPQM Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 65%.

The sperm-specific protein Izumo, named for a Japanese shrine dedicated to marriage, is essential for sperm-egg plasma membrane binding and fusion.The sperm-specific protein Izumo, named for a Japanese shrine dedicated to marriage, is essential for sperm-egg plasma membrane binding and fusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IZUMO1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Izumo1
Gene Accession No. Q8IYV9
Gene Alias 1700058F15Rik; Izumo; izumo sperm-egg fusion 1; izumo sperm-egg fusion protein 1; IZUMO1; MGC34799; OBF; oocyte binding/fusion factor; Sperm-specific protein izumo
Gene Symbols Izumo1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IZUMO1. Recombinant protein control fragment (Product #RP-90522).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284359
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.