Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IZUMO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569973 Supplier Invitrogen™ Supplier No. PA569973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 83%; Pig: 86%; Rat: 83%.

IZUMO4 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IZUMO4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IZUMO4
Gene Accession No. Q1ZYL8
Gene Alias 9030607L17Rik; AI550407; C19orf36; IMAGE:4215339; IZUMO family member 4; izumo sperm-egg fusion protein 4; Izumo4; putative SAMP14 binding protein variant 1; putative SAMP14 binding protein variant 2; RGD1564722; sperm 22 kDa protein c113; UNQ831/PRO1758
Gene Symbols IZUMO4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human IZUMO4 (aa 183-232).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113177
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.