Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ J 104129 fumarate
GREENER_CHOICESDP

Catalog No. 250710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-071-0 Supplier Tocris Bioscience™ Supplier No. 2507/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective M3 antagonist

Potent M3 muscarinic receptor antagonist that displays ∽ 120-fold selectivity over M2 receptors (Ki values are 4.2, 19 and 490 nM for human M3, M1 and M2 receptors respectively). Exhibits > 250-fold bronchial selectivity; inhibits ACh-induced bronchoconstriction but not ACh-induced bradycardia (KB values are 3.3 and 170 nM for rat trachea M3 and rat right atria M2 receptors respectively).

Chemical Identifiers

CAS 257603-40-0
Molecular Formula C28H40N2O6

Specifications

Quantity 10 mg
Formula Weight 500.63
Purity >99%
Molecular Weight (g/mol) 500.636
Chemical Name or Material J 104129 fumarate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.