Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (±)-J 113397
GREENER_CHOICESDP

Catalog No. 259810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-981-0 10 mg
25-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-981-0 Supplier Tocris Bioscience™ Supplier No. 2598/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NOP antagonist

Potent and selective NOP receptor antagonist (IC50 values are 2.3, 1400, 2200 and > 10000 nM for NOP, κ, μ and δ-opioid receptors respectively). Inhibits nociceptin/orphanin FQ-induced hyperalgesia in the mouse tail-flick test.

Chemical Identifiers

CAS 217461-40-0
Molecular Formula C24H37N3O2

Specifications

Quantity 10 mg
Formula Weight 399.57
Purity >98%
Molecular Weight (g/mol) 399.579
Chemical Name or Material (±)-J 113397
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.