Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ J 147
GREENER_CHOICESDP

Catalog No. 508110R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
508110R 10 mg
508150R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 508110R Supplier Tocris Bioscience™ Supplier No. 5081/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Neuroprotective and neurotrophic compound; reduces Aβ40 and Aβ42 levels

J 147 is a potent neuroprotective and neurotrophic compound. Protects against neurotoxicity in cortical neurons in vitro (EC50 = 25 - 200 nM). Reduces soluble Aβ40 and Aβ42 levels and increases BDNF levels in the hippocampus in vivo. Enhances memory in wild type rats. Prevents memory deficits and rescues cognitive deficits in young and aged transgenic Alzheimer's disease mice, respectively. Orally active.

Specifications

Quantity 10 mg
Product Type J 147
Purity >99%
Molecular Weight (g/mol) 350.33
Inhibitors J 147

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.