Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ J Chain Recombinant Rabbit Monoclonal Antibody (PD00-89)
GREENER_CHOICE

Catalog No. FEMA550730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550730 Supplier Invitrogen™ Supplier No. MA550730
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Human tonsil tissue, Daudi. Predicted band size: 18 kDa Subcellular Location: Secreted.

J chain is a small glycopeptide and is structurally unrelated to heavy or light chains, but is synthesized by all plasma cells that secrete polymeric immunoglobulins. J chain is linked to IgA and IgM by disulfide bonds, however, it has been detected in IgG- and IgD-containing cells. J chains are present in a large proportion of the immunoglobulin-positive cells in the germinal centers of the tonsil and lymph node. B cells secrete J chain at an early stage of differentiation with the expression persisting in those cells destined to produce IgA or IgM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen J Chain
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD00-89
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene JCHAIN
Gene Accession No. P01591
Gene Alias 9530090F24Rik; AI323815; IGCJ; Igj; IgJ chain; immunoglobulin J chain; immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides; immunoglobulin joining chain; immunoglobulin joining gene (not J region); J chain; Jch; JCHAIN; Joining chain of multimeric IgA and IgM
Gene Symbols JCHAIN
Host Species Rabbit
Immunogen Synthetic peptide within Human J Chain aa 50-150 (internal sequence).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3512
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.