Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAB1 Antibody (OTI2B12), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP27169A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27169A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27169A70 Supplier Novus Biologicals Supplier No. NBP271659AF750
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JAB1 Monoclonal specifically detects JAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen JAB1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2B12
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias COP9 constitutive photomorphogenic homolog subunit 5 (Arabidopsis), COP9 signalosome complex subunit 5, CSN5COP9 (constitutive photomorphogenic, Arabidopsis, homolog) subunit 5, EC 3.4, JAB138 kDa Mov34 homolog, Jun activation domain-binding protein 1, MOV-34, SGN5MGC3149, Signalosome subunit 5
Gene Symbols COPS5
Host Species Mouse
Immunogen Full length recombinant protein of human JAB1 (NP_006828) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10987
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.